Learn More
Memmert Pass-through Oven UF160TS TwinDISPLAY, 161L, 20-250°C, Forced Convection
Brand:  Memmert UF160TS
Pass-through Oven UF160TS TwinDISPLAY, 161L, 20-250°C, Forced Convection
Regulatory: This product is under eco-contribution tax.
Product Code. 16388027
  • ONLINE EXCLUSIVE PRICE
    9200.00€
    8917.00€ / Each
    Save 283.00€
    3% Off
Only null left
Estimated Shipment: 03-02-2026