missing translation for 'onlineSavingsMsg'
Learn More

Checkers Park-It™, Black/White

The rubber Park-It™ Parking Curb is used to effectively guide cars when pulling into a parking place. It is profiled at 10 cm high, safeguarding potential damage to today’s low, front-end car designs. They are more resistant than concrete or plastic curbs.

44.00€ - 74.00€

Specifications

Color Black, White
Material Recycled Rubber
Width (Metric) 15 cm
View More Specs

Products 3
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
17712318
View Documents
Checkers
GNRS1320WB
90 cm
44.00€
Each
Estimated Shipment: 03-12-2024
to see stock.
Add to basket
 
17732318
View Documents
Checkers
GNRS1420WB
120 cm
57.00€
Each
Estimated Shipment: 03-12-2024
to see stock.
Add to basket
 
17752318
View Documents
Checkers
GNRS1620WB
180 cm
74.00€
Each
Estimated Shipment: 03-12-2024
to see stock.
Add to basket
 
Description

Description

  • The Park-It™ parking curb is constructed of 100% recycled rubber and is used to effectively guide cars when pulling into a parking stall
  • It is profiled at 10 cm high, safeguarding potential damage to today's low, front-end car designs
  • Unlike concrete or plastic, which necessitate frequent replacements, the Park-It™'s recycled rubber construction makes it safer and more durable
  • In-moulded highway reflective tape (available in yellow or white) ensures visibility and pedestrian safety
  • The Park It™ comes with pre-moulded holes and can be shipped with rebar spikes or lag bolts for installation
  • They are lightweight (16 kg for an 18 cm Park-It™) and flexible; conforming to the contour of uneven road surfaces
  • Park-It™ parking curbs are available in 90 cm, 120 cm and 180 cm lengths and are covered by a 3-year warranty; 1 year on the tape (under normal use)
Specifications

Specifications

Black, White
Recycled Rubber
15 cm
Videos
SDS
Documents

Documents

Product Certifications