missing translation for 'onlineSavingsMsg'
Learn More

Bemis™ Parafilm™ M Laboratory Wrapping Film

Efficient and cost-effective covering for test tubes, beakers, vials, petri dishes, flasks, and various other instruments and objects with irregular surfaces.

Brand:  Bemis™ PM996

 View more versions of this product

Product Code. 12613586

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Efficient and cost-effective covering for test tubes, beakers, vials, petri dishes, flasks, and various other instruments and objects with irregular surfaces.

  • Protect the integrity of research applications with a tight moisture-barrier that holds a strong seal and doesn’t release from glassware, despite off-gassing
  • Reduce the risk of tampered data with a covering that is odorless, colorless and semi-transparent
  • Prevents moisture and volume loss due to material composition and tight seal
  • Flexible and self-sealing material allows it to cling around irregular shapes and surfaces
  • Stretches more than 200% of original length
  • Resistant up to 48 hours against many polar substances such as saline solutions, inorganic acids and alkaline solutions

Permeability

  • Oxygen: (ASTM 1927-98) 150 cc/m² d at 23°C and 50% RH
  • Carbon Dioxide: (Modulated IR Method) 1200 cc/m² d at 23°C and 0% RH
  • Water Vapor: (ASTM F1249-01)
    • Flat: 1 g/m² d at 38°C and 90% RH
    • Creased: 1 g/m² d at 38°C and 90% RH

Effects of Common Reagents

  • Potassium Permanganate
    • 5%: No apparent effect except permanent dark brown coloration in 18 hours
    • 0.1%: Same as 5% except slightly less color
  • Iodine solution (0.1N)
    • No effect except staining brown in 18 hours
  • Ethyl Alcohol (95%)
    • No apparent effect except some face whitening in 24 hours
  • No apparent effect in 24 hours:
    • Hydrochloric Acid conc. (12N) dil. (5N)
    • Sulphuric Acid conc. (36N) dil. (5N)
    • Nitric Acid conc. (16N) dil. (5N)
    • Sodium Hydroxide conc. (22%)
    • Ammonium Hydroxide conc. (28% NH<sub>3</sub>)
    • Salt (NaCl) solution (20%)
    • Isopropyl Alcohol (99%)
  • Not recommended for use with chlorinated, non-polar aliphatic and aromatic solvents
  • Film becomes soft and sticky at about 130° to 150°F (54° to 66°C)

Order Info

13-374-10, 13-374-12, and 13-374-16 rolls are packaged in cardboard dispensing cartons.

Specifications

Specifications

Efficient and cost-effective covering, over wrap to further secure stoppers, lids, and caps, and shelf or tray liner to prevent slipping of bottles or instruments.
Sealing Film
10 cm
Thermoplastic
125 ft.
4 in.
38 m
Parafilm Wrap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions