missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Legionella MWY Selective Supplement
Oxoid Legionella MWY Selective Supplement is used for the isolation of Legionella pneumophila from potable water samples.
201.00€ - 656.88€
Specifications
Species | Legionella |
---|---|
Description | Legionella MWY Selective Supplement |
Form | Freeze-dried |
Product Type | Selective Supplement |
Quantity | 10/Pk. |
Product Code | Brand | Yield | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Yield | Price | Quantity & Availability | |||||
17140671
|
Thermo Scientific™
SR0255B |
Each vial for 500mL medium |
656.88€
Pack of 10 |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||||
17150671
|
Thermo Scientific™
SR0255E |
Each vial for 100mL medium |
201.00€
Pack of 10 |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||||
Description
Oxoid Legionella MWY Selective Supplement is used for the isolation of Legionella pneumophila from potable water samples.
Add to Legionella Agar Base, Part No. CM1203BSpecifications
Legionella | |
Freeze-dried | |
10/Pk. |
Legionella MWY Selective Supplement | |
Selective Supplement |