Learn More
WTW™ OxiTop™ IS 12 Measuring Head
Respirometric OxiTop™ measuring system for BOD self check measurement with undiluted samples – measuring ranges up to 4,000 mg/L BOD possible
Brand: WTW™ 208211
This item is not returnable.
View return policy
Description
- Easy handling without dilution series
- Data security by built-in measured value memory
- Suitabe for immediate use
Specifications
BOD bottles with amber, magnetic bars, bar puller, sodium hydroxide pellets, NaOH tanks, volumetric flasks 164 and 432mL. | |
CE | |
1 Year | |
42 cm | |
±1% | |
100/240 V |
Measurement using OxiTop™ is based on pressure measurement in a closed system | |
11 kg | |
5°C to 50°C | |
28 cm | |
100/240V, 50/60Hz | |
50/60 Hz |