Learn More
WTW™ OxiTop™ Control B6M Package
Brand:  WTW™ 209334
212.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10293721
  • 224.00€ / Each
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket
Specifications
OxiTop™ Control B6M Package
Laboratory method for determining the microbial soil respiration
6 MG 1 measuring vessels, 1000mL, stopper adapters for OxiTop™-C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
WTW™ OxiTop™ Control B6M Package > 6 measuring units; 6mg vessels