Learn More
WTW™ Oxi 3210 e Waterproof Dissolved Oxygen Meter
Brand: WTW™ 2BA200
Specifications
Automatic from 500 to 1100 mbar | |
0.0 to 200, 0 to 1250mbar | |
±0.1°C | |
CE | |
7 x 3.12 x 2.25in. | |
3 Years | |
0.1, 1 | |
0.00 to 90mg/L | |
Manual input, automatic correction from 0 to 70 ppt | |
Up to 1000hr. | |
18 cm | |
5.5 cm |
For challenging measurements in the field and plant, e.g. water/wastewater analysis, chemical/pharmaceutical industry, etc | |
0.1°C | |
Backlit Graphic LCD | |
IP-66/67 | |
0.6 kg | |
4 x 1.5V AA or 4 x 1.2V NiMH rechargeable batteries | |
Galvanic | |
0.01, 0.1mg/L | |
-5°C to +50°C | |
Four AA | |
8 cm | |
±0.5% |