missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OXA1L Mouse anti-Human, Clone: 4A2D9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66128-1-IG-20UL
This item is not returnable.
View return policy
Description
OXA1L is required for the insertion of integral membrane proteins into the mitochondrial inner membrane. It is essential for the activity and assembly of cytochrome oxidase and is required for the correct biogenesis of ATP synthase and complex I in mitochondria.Specifications
| OXA1L | |
| Monoclonal | |
| 0.82 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15070 | |
| OXA1L | |
| OXA1L Fusion Protein Ag13819 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 4A2D9 | |
| Unconjugated | |
| OXA1L | |
| Hsa, OXA1 like protein, OXA1Hs, OXA1L | |
| Mouse | |
| Protein G | |
| RUO | |
| 5018 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction