missing translation for 'onlineSavingsMsg'
Learn More

OXA1L Mouse anti-Human, Clone: 4A2D9, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66128-1-IG-150UL

 View more versions of this product

Product Code. 16827725

  • 410.00€ / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

OXA1L is required for the insertion of integral membrane proteins into the mitochondrial inner membrane. It is essential for the activity and assembly of cytochrome oxidase and is required for the correct biogenesis of ATP synthase and complex I in mitochondria.
Specifications

Specifications

OXA1L
Monoclonal
0.82 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q15070
OXA1L
OXA1L Fusion Protein Ag13819
150 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
4A2D9
Unconjugated
OXA1L
Hsa, OXA1 like protein, OXA1Hs, OXA1L
Mouse
Protein G
RUO
5018
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.