Learn More
Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System Combs
missing translation for 'orderingAttributeHoverText'
Teeth Count:
17
Thickness (Metric) Teeth:
1 mm
1.5 mm
missing translation for 'unitSize'
Each
Beschreibung
Eliminate comb assembly with Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System Combs. These heavy-duty, one-piece microwell combs are for use with the Owl D4 Electrophoresis System.
Heavy-duty, one-piece combs eliminate comb assembly.
Ordering Information:
Customized combs are available (Order SKU # XCM). Please contact your Customer Service or Sales Representative for further details.
Spezifikation
Spezifikation
| Description | Micro well comb |
| For Use With (Equipment) | Owl™ D4 Electrophoresis System |
| Product Line | Owl |
| Type | Comb |
| Teeth Count | 17 |
| Thickness (Metric) Teeth | 1.5 mm |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur