Learn More
Osmolality Standard, 300mOsm/kg H2O, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
12 x 5 mL
Specifications
Specifications
Packaging | Ampoules |
Quantity | 12 x 5 mL |
Physical Form | Liquid |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
OSM30024H1 | Certificate of Analysis | 18/02/2025 | RE-OSM-300 |
OSM30024C1 | Certificate of Analysis | 08/09/2024 | RE-OSM-300 |
OSM30024B1 | Certificate of Analysis | 07/08/2024 | RE-OSM-300 |
OSM30023C1 | Certificate of Analysis | 17/01/2024 | RE-OSM-300 |
OSM30022M1 | Certificate of Analysis | 23/05/2023 | RE-OSM-300 |
Product Content Correction
Product Title
Osmolality Standard, 300mOsm/kg H2O, Reagecon™ >