missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ornithine Decarboxylase Antibody (ODC1/487), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47837AF488
This item is not returnable.
View return policy
Description
Ornithine Decarboxylase Monoclonal antibody specifically detects Ornithine Decarboxylase in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
Specifications
Ornithine Decarboxylase | |
Monoclonal | |
Alexa Fluor 488 | |
ODCEC 4.1.1.17, ornithine decarboxylase, ornithine decarboxylase 1 | |
Recombinant human Ornithine Decarboxylase protein (Uniprot: P11926) | |
0.1 mL | |
Cell Cycle and Replication | |
4953 | |
Store at 4°C in the dark. | |
IgG2a κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
ODC1/487 | |
50 mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ornithine Decarboxylase Antibody (ODC1/487), Alexa Fluor™ 488, Novus Biologicals™ > 0.1 mL, Alexa Fluor 488
Spot an opportunity for improvement?Share a Content Correction