Learn More
Thermo Scientific™ Orion™ Redox/ORP/Temp Electrodes
Product Code. 10315173
Click to view available options
Type:
Epoxy Refillable Triode
Epoxy Triode Low Maintenance
For Use With (Equipment):
Orion A+ Series Meters
Orion Star Series Meters
Unit Size:
Each
Product Code. 10315173
Brand: Thermo Scientific™ 9180BN
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Accurately measure ORP and temperature together in just one electrode, with a built-in temperature sensor.
Thermo Scientific™ Orion™ Triode 3-in-1 pH/Automatic Temperature Compensation Electrodes come with a built-in ATC and are engineered with an epoxy body for ruggedness. The 9179BN and 9179BNMD are low-maintenance gel-filled electrodes, best suited for educational institutes where minimal maintenance is required, while the 9180BNMD is a refillable electrode best suited for general purpose samples.
Features:
  • Available in epoxy triode low maintenance and epoxy refillable triode construction
  • Built-in thermistor for Automatic Temperature Compensation (ATC)
  • Not designed for amperometric or constant current titrations, such as Karl Fischer analysis
TRUSTED_SUSTAINABILITY
Specifications
Description Epoxy Refillable Triode
Material Epoxy
For Use With (Equipment) Orion A+ Series Meters
Type Epoxy Refillable Triode