Learn More
Thermo Scientific™ Orion™ ISE Surfactant Analysis Solutions
Use Thermo Scientific™ Orion™ ISE Surfactant Analysis Solutions for titrations with surfactant ion selective electrodes (ISE).
Brand:  Thermo Scientific™ 654201
 View more versions of this product
Product Code. 12879201
  • 81.10€ / 475mL
Explore more special offers
Description
Description
Use Thermo Scientific™ Orion™ ISE Surfactant Analysis Solutions for titrations with surfactant ion selective electrodes (ISE).
All surfactant analysis solutions are packaged as one 1 pint (475mL) bottle, except Cat. No. 654202 packaged as one 2 oz. (60mL) bottle.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hyamine 1622 Titrant, 475mL
475 mL
Bottle
Promotions
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.