Learn More
Thermo Scientific™ Torches for iCAP™ Q/Qnova Series ICP-MS Systems
Get improved plasma stability from the tulip design and easy, push-in insertion of Thermo Scientific™ Torches for iCAP™ Q/Qnova Series ICP-MS systems.
351.00€
Specifications
Type | ICP-MS Torch |
---|---|
For Use With (Equipment) | iCAP Q/Qnova Series ICP-MS Systems |
Product Code | Brand | Type | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Description | Price | Quantity & Availability | ||||
13552790
|
Thermo Scientific™
1230790 |
ICP-MS Torch | Quartz Torch |
351.00€
Each |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Gain improved plasma stability from the tulip design and easy, push-in insertion of fully demountable, single piece, Thermo Scientific™ Torches with connection-free gas supply for Thermo Scientific™ iCAP™ Q/Qnova Series ICP-MS systems. All connectivity (argon gas supplies and plasma ignition) is contained within the torch holder to reduce complexity and improve usability.
Specifications
ICP-MS Torch | |
iCAP Q/Qnova Series ICP-MS Systems |