Learn More
3M™ Orange SMS Based Fabric Category III Protective Coveralls
Product Code. p-8003857
Click to view available options
Size:
Small
Large
Unit Size:
Pack of 20
Product Code. 11933754
Brand: 3M™ 7000089634
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Specifications
Product Type General Purpose Coveralls
Size Large
Gender Unisex
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 2-panel
Seam Type Serged
Sterility Non-sterile
Protection Class Category III
Height (Metric) 174 to 181 cm