Learn More
Tango™ OPRL1-bla U2OS Cells
Tango™ OPRL1-bla U2OS Cells contain the human Opioid Receptor-Like 1 (OPRL1) linked to a TEV protease site.
Brand:  Tango™ K1786
Additional Details : Weight : 0.01000kg
Product Code. 10448073
  • 12500.00€ / Each
Estimated Shipment: 23-07-2024
to see stock.
Add to basket
Description
Description
  • Gene Symbol: OPRL1
  • Accession Number: NM_000913
  • Cytoplasmic Tail: Tail from AVPR2 GPCR
  • Population Status: Single Cell Clone Origin
  • The Tango™ OPRL1-bla U2OS cells contain the human Opioid Receptor-Like 1 (OPRL1) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.
  • The Tango™ OPRL1-bla U2OS cells have been functionally validated for Z' factor and EC50 concentrations of established ligands. In addition, Tango™ OPRL1-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary for each product.
Specifications
Specifications
Liquid Nitrogen
GPCRs
8 x 106 Cells
Fluorescent
OPRL1-bla U2OS cells, OPRL1-bla U2OS Assay Protocol, Certificate of Analysis
Dividing Cells