missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Dibafit™ Unions with 1/4 in. 28 UNF(F) Connection

Easily connect 1/4 in. 28 UNF(M) fittings with compatible unions

Brand:  Cole-Parmer™ 21939-96

Product Code. 17102445

  • 38.50€ / Pack of 10
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Available in polypropylene (PP), ETFE, or PEEK materials
Specifications

Specifications

UNF Threaded Coupling
7.62 cm
Chromatography
10/Pk.
1.27 cm
Polypropylene
0.25 in. to 28 two way flat bottom
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions