missing translation for 'onlineSavingsMsg'
Learn More

Oligomycin, mixture of A, B and C, from Streptomyces diastatochromogenes, ACROS Organics™

Catalog Number 10154950
Click to view available options
5mg; Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy
Type Oligomycin, Mixture of A, B and C
Physical Form Powder
CAS 1404-19-9
Infrared Spectrum Authentic
Molecular Formula C45H74O11
MDL Number MFCD00065705,MFCD01779388
Packaging Glass Bottle
InChI Key MNULEGDCPYONBU-ZUSSGZTJNA-N
SMILES CCC1CCC2OC3(CCC(C)C(CC(C)O)O3)C(C)C(OC(=O)\C=C\C(C)C(O)C(C)C(=O)C(C)C(O)C(C)C(=O)C(C)(O)C(O)C(C)C\C=C\C=C/1)C2C
IUPAC Name (4E,18E,20Z)-22-ethyl-7,11,14,15-tetrahydroxy-6'-(2-hydroxypropyl)-5',6,8,10,12,14,16,28,29-nonamethyl-2,26-dioxaspiro[bicyclo[23.3.1]nonacosane-27,2'-oxane]-4,18,20-triene-3,9,13-trione
Molecular Weight (g/mol) 791.08
PubChem CID 78358496
Formula Weight 791.08
Percent Purity ≥92% (Total oligomycin A, B, C)
Color White to Yellow
Quantity 5 mg
Chemical Name or Material Oligomycin, mixture of A, B and C, From Streptomyces Diastatochromogenes
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'