Learn More
OHAUS™ Sample Pan for MB Series Moisture Analyzers
Product Code. p-3207089 Shop All Ohaus Products
Click to view available options
Type:
Disposable Sample Pan
Reusable Sample Pan
Quantity:
3/Pk.
80/Pk.
Unit Size:
Pack of 3
Pack of 80
Product Code. 11804242
Brand: OHAUS™ 80850086
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Sample Pans for OHAUS™ MB series moisture analyzers.
  • Shape: round
  • Choose disposable or reusable
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) US: MB35,MB45,MB27,MB90,MB120 Moisture Analyzers
EU: MB27, MB25, MB90-GB, MB120-GB, MB23, MB120-EU
Size 101.6 mm diameter
Quantity 80/Pk.
Type Disposable Sample Pan