missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Octanoylcarnitine chloride, Tocris Bioscience™
Click to view available options
Quantity:
50 mg
Description
Homolog of acetylcarnitine chloride (Cat. No. 0355). Acylcarnitines are important intermediates in lipid metabolism.
Specifications
Specifications
CAS | 18822-86-1 |
Molecular Formula | C15H30ClNO4 |
Quantity | 50 mg |
Synonym | octanoyl-carnitine, octanoyl carnitine hydrochloride, +/--octanoylcarnitine chloride, octanoylcarnitine chloride, rac octanoyl carnitine chloride, octanoyl-dl-carnitine hydrochloride, 3-carboxy-2-octanoyloxy propyl trimethylazanium chloride, octanoyl-dl-carnitin chloride, a+/--octanoylcarnitine chloride, octanoic acid, ester with 3-carboxy-2-hydroxypropyl trimethylammonium chloride |
InChI Key | MYMFUYYXIJGKKU-UHFFFAOYSA-N |
SMILES | CCCCCCCC(=O)OC(CC(=O)O)C[N+](C)(C)C.[Cl-] |
IUPAC Name | (3-carboxy-2-octanoyloxypropyl)-trimethylazanium;chloride |
Molecular Weight (g/mol) | 323.858 |
PubChem CID | 177508 |
Formula Weight | 323.86 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction