Learn More
Ochratoxin A-(phenyl-d5)-Lösung, Honeywell Fluka™
50 μg/mL in acetonitrile, analytical standard, reference material
Brand: Honeywell Fluka R1324-2ML
This item is not returnable.
View return policy
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
- Free expert technical support
Specifications
75-05-8, 666236-28-8 | |
2 mL | |
0.78 g/cm3 | |
6°C | |
Liquid |
Glass Bottle | |
Analytical Standard | |
82°C | |
Mycotoxins |