Learn More
DWK Life Sciences O-ring, for stopcock keys with groove
For securing the DURAN™ valve plug with groove in the DURAN™ single way stopcock.
Brand:  DWK Life Sciences 286031408
 View more versions of this product
Product Code. 15991501
  • 28.55€ / Pack of 50
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Description
The O-ring in the groove of the DURAN™ tap plug provided for this purpose secures the tap plug in the disposable tap.
  • Secures the valve plug from falling out of the valve
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sealing
24.0 NS
100/Pk.