missing translation for 'onlineSavingsMsg'
Learn More

o-Dichlorobenzene, HPLC Grade, J.T. Baker™

Product Code. 15369034
Click to view available options
Quantity:
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

High purity solvents for use in analytical and preparative separations

Specifications

CAS 95-50-1
Flash Point 66.00°C
Packaging Narrow Mouth Amber Glass Bottle
Molecular Formula C6H4Cl2
Quantity 4 L
Specific Gravity 1L = 1.30kg
Synonym o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben
InChI Key RFFLAFLAYFXFSW-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)Cl)Cl
IUPAC Name 1,2-dichlorobenzene
Molecular Weight (g/mol) 146.998
PubChem CID 7239
ChEBI CHEBI:35290
Formula Weight 147
Grade Meets USP requirements
Chemical Name or Material o-Dichlorobenzene, Bulk Pharmaceutical Chemical
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'