Learn More
B2M™ Nylon Nail Brush
Designed to clean nails, hands and forearms. B2M™ Nylon Nail Brush has hard nylon bristles to remove debris and microorganisms in order to prevent the spread of biological contaminants and infectious disease. It can be autoclaved for effective sterilization.
Brand:  B2M™ 40294
Product Code. 11738602
  • 23.00€ / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Thick handle fits easily in the palm
  • Flat brush with highly resistant nylon bristles
  • Can be autoclaved for use in sterile environments
Specifications
Specifications
Nail Brush
Autoclavable
Cleaning of nails, hands and forearms
Nylon
White
1 Ea.