Learn More
Masterflex™ Nylon Barbed Fitting Reducing Union
Click to view available options
For Use With:
115.87 x 12.7mm
12.7 x 12.7mm
19.05 x 12.7mm
6.35 x 3.175mm
Unit Size:
Pack of 10
Specifications
Specifications
| Material | Nylon |
| For Use With | 19.05 x 12.7mm |
| Min. Operating Temperature (Metric) | -46°C |
| Max. Operating Temperature (Metric) | 121°C |
| Product Type | Barbed Fitting Reducing Union |