Learn More
Masterflex™ Nylon Barbed Fitting Reducing Union
Product Code. 12427260
Click to view available options
For Use With:
115.87 x 12.7mm
12.7 x 12.7mm
19.05 x 12.7mm
6.35 x 3.175mm
Unit Size:
Pack of 10
Product Code. 12427260
Brand: Masterflex™ 3061614
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Specifications
Material Nylon
For Use With 6.35 x 3.175mm
Min. Operating Temperature (Metric) -46°C
Packaging Quantity 10 Pack
Max. Operating Temperature (Metric) 121°C
Product Type Barbed Fitting Reducing Union