Learn More
Tocris Bioscience™ NVP ADW 742
ATP-competitive inhibitor of IGF1R
Brand:  Tocris Bioscience™ 5247/10
152.91 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18753968
  • 173.00€ / 10mg
Explore more special offers
Description
Description
NVP ADW 742 is an ATP-competitive inhibitor of IGF1R (IC50 in the range of 0.1-0.2 μM). Also inhibits c-Kit kinase activity (IC50 in the range of 3-5 μM). Exhibits a synergistic effect with Imatinib (Cat. No. 5906), enhancing the sensitivity of multiple small cell lung cancer cell lines to the chemotherapeutics Etoposide (Cat. No. 1226) and Carboplatin (Cat. No. 2626).
Specifications
Specifications
>99%
NVP ADW 742
NVP ADW 742
10 mg
453.58