Learn More
Thermo Scientific™ Nunc™ Cryobank and Bank-It™ Freezer Rack
Click to view available options
Color:
Silver
Unit Size:
Each
Description
Hold nineteen 0.5mL cryobank racks or thirteen 1.0mL cryobank racks with the Thermo Scientific™ Nunc™ Cryobank and Bank-It™ Vial Rack.
This verticle aluminum freezer rack with 74 cm in height is specifically designed to hold Thermo Scientific Nunc Cryobank in lock racks.
Specifications
Specifications
| Description | Cryobank and Bank-It Vial Rack |
| Capacity (Metric) | 0.5 mL (13 Cryobank Rack), 1 mL (13 Cryobank Rack) |
| Holds | 19 x 0.5mL or 13 x 1.0mL cryobank racks; one each |
| Material | Aluminum |
| Type | Cryobank and Bank-It Vial Rack |
| Coding | 2D Barcode |
| Vial Type | Cryobank |
| Color | Silver |
| For Use With (Equipment) | 19 x 0.5mL or 13 x 1.0mL cryobank racks |
Product Content Correction
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto