missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSE2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA544399
This item is not returnable.
View return policy
Description
Peptide sequence: KFLALQSKNS DADFQNNEKF VQFKQQLKEL KKQCGLQADR EADGTEGVDE Sequence homology: Cow: 86%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 79%; Rabbit: 100%
NSE2 Polyclonal antibody specifically detects NSE2 in Human samples. It is validated for Western Blot.Specifications
NSE2 | |
Polyclonal | |
Unconjugated | |
Nsmce2 | |
1110014D18Rik; AI661537; C8orf36; E3 SUMO-protein ligase NSE2; E3 SUMO-protein transferase NSE2; FLJ32440; hMMS21; methyl methanesulfonate sensitivity gene 21; MMS21; MMS21 homolog; non-SMC element 2 homolog; non-SMC element 2 homolog (MMS21, S. cerevisiae); non-SMC element 2, MMS21 homolog; non-SMC element 2, MMS21 homolog (S. cerevisiae); non-structural maintenance of chromosomes element 2 homolog; NSE2; NSE2 (MMS21) homolog, SMC5-SMC6 complex SUMO ligase; NSE2/MMS21 homolog, SMC5-SMC6 complex SUMO ligase; NSMCE2; RGD1305156; Unknown (protein for MGC:133996); zinc finger, MIZ-type containing 7; ZMIZ7 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
286053 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96MF7 | |
Nsmce2 | |
Synthetic peptide directed towards the middle region of human NSMCE2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |