missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSE2 Monoclonal Antibody (OTI4A2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA525224
This item is not returnable.
View return policy
Description
This gene encodes the large subunit of DNA damage-binding protein which is a heterodimer composed of a large and a small subunit. This protein functions in nucleotide-excision repair. Its defective activity causes the repair defect in the patients with xeroderma pigmentosum complementation group E (XPE). However, it remains for mutation analysis to demonstrate whether the defect in XPE patients is in this gene or the gene encoding the small subunit. In addition, Best vitelliform mascular dystrophy is mapped to the same region as this gene on 11q, but no sequence alternations of this gene are demonstrated in Best disease patients.Specifications
NSE2 | |
Monoclonal | |
0.49 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96MF7 | |
Nsmce2 | |
Full length human recombinant protein of NSMCE2 produced in E.coli | |
100μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Western Blot | |
OTI4A2 | |
Unconjugated | |
Nsmce2 | |
1110014D18Rik; AI661537; C8orf36; E3 SUMO-protein ligase NSE2; E3 SUMO-protein transferase NSE2; FLJ32440; hMMS21; methyl methanesulfonate sensitivity gene 21; MMS21; MMS21 homolog; non-SMC element 2 homolog; non-SMC element 2 homolog (MMS21, S. cerevisiae); non-SMC element 2, MMS21 homolog; non-SMC element 2, MMS21 homolog (S. cerevisiae); non-structural maintenance of chromosomes element 2 homolog; NSE2; NSE2 (MMS21) homolog, SMC5-SMC6 complex SUMO ligase; NSE2/MMS21 homolog, SMC5-SMC6 complex SUMO ligase; NSMCE2; RGD1305156; Unknown (protein for MGC:133996); zinc finger, MIZ-type containing 7; ZMIZ7 | |
Mouse | |
Affinity Chromatography | |
RUO | |
286053 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |