missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSC 663284, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
Potent, selective inhibitor of Cdc25 dual specificity phosphatases (Ki values are 29, 95 and 89 nM for human Cdc25A, Cdc25B2 and Cdc25C respectively); > 20- and > 450-fold selective over VHR and PTP1B phosphatases respectively. Arrests cells at both G1 and G2/M phase and blocks cdk2 and cdk1 activation. Blocks proliferation of a range of human tumor cell lines (IC50 = 0.2 - 35 μM).
Specifications
Specifications
CAS | 383907-43-5 |
Molecular Formula | C15H16ClN3O3 |
MDL Number | MFCD08276924 |
Quantity | 10 mg |
Synonym | cdc25 phosphatase inhibitor ii, nsc 663284, 6-chloro-7-2-morpholin-4-yl-ethylamino quinoline-5,8-dione, 6-chloro-7-2-morpholin-4-yl ethyl amino quinoline-5,8-dione, d0xu7m, nsc hplc , solid, 5,6-chloro-7-2-4-morpholinyl ethyl amino, cdc25 phosphatase inhibitor ii, nsc 663284_x000d_, 6-chloro-7-2-morpholinoethyl amino quinoline-5,8-dione, 7-n-aminoethyl-4-morpholino-6-chloro-5,8-quinolinedione, 6-chloro-7-2-morpholin-4-ylethylamino quinoline-5,8-dione |
InChI Key | BMKPVDQDJQWBPD-UHFFFAOYSA-N |
SMILES | ClC1=C(NCCN2CCOCC2)C(=O)C2=C(C=CC=N2)C1=O |
IUPAC Name | 6-chloro-7-{[2-(morpholin-4-yl)ethyl]amino}-5,8-dihydroquinoline-5,8-dione |
Molecular Weight (g/mol) | 321.76 |
PubChem CID | 379077 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'