missing translation for 'onlineSavingsMsg'
Learn More

NPS 2390, Tocris Bioscience™

Product Code. 18065452
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Group I mGlu antagonist

Group I mGlu antagonist; displays noncompetitive antagonist activity at both mGlu1 and mGlu5 receptors. Thought to act on a site separate from the glutamate binding pocket.

Chemical Identifiers

CAS 226878-01-9
Molecular Formula C19H21N3O
Molecular Weight (g/mol) 307.397
InChI Key ZKFVOZCCAXQXBU-UHFFFAOYSA-N
Synonym n-adamantan-1-yl quinoxaline-2-carboxamide, n-1-adamantyl quinoxaline-2-carboxamide, quinoxaline-2-carboxylic acid adamantan-1-ylamide, n-tricyclo 3.3.1.13,7 dec-1-yl-2-quinoxalinecarboxamide, d09cpy, d0w0xm, n-1-adamantyl-2-quinoxalinecarboxamide, nps hplc , solid, 2-quinoxalinecarboxamide, n-tricyclo 3.3.1.13,7 dec-1-yl, n-3r,5s,7s-adamantan-1-yl quinoxaline-2-carboxamide
PubChem CID 7067728
IUPAC Name N-(1-adamantyl)quinoxaline-2-carboxamide
SMILES C1C2CC3CC1CC(C2)(C3)NC(=O)C4=NC5=CC=CC=C5N=C4

Specifications

Quantity 10 mg
Formula Weight 307.39
Percent Purity >99%
Chemical Name or Material NPS 2390
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'