missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Dicer knockout Mouse Embryonic Stem Cells

Used in common research applications

Brand:  Novus Biologicals™ NBP1-96751

Product Code. 18755673

  • 646.00€ / 2mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Cells are supplied in 2mL quantities (about 1 x 106 cells/mL) in freezing media (60% DMEM, 20% FBS, 20% DMSO)
  • Gene ID: 23405
  • Gene symbol: DICER1

Recommended Storage

Store in gas phase of liquid nitrogen

Services

6 Months

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in gas phase of liquid nitrogen
Dicer knockout Mouse embryonic stem cells
2mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Novus Biologicals™ Dicer knockout Mouse Embryonic Stem Cells > Dicer knockout Mouse Embryonic Stem Cells

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

This product is for research use only and is not approved for use in humans or in clinical diagnosis