missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ beta 2-Microglobulin Protein

Highly purified. Generating reliable and reproducible results.

403.00€ - 1025.00€

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 20% glycerol, 2mM DTT, 100mM NaCl
Molecular Weight (g/mol) 14.0kDa
Purification Method SDS-PAGE
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18229482
View Documents
Novus Biologicals™
NBP1-50840-0.05MG
0.05 mg
1025.00€
0.05mg
Please to purchase this item. Need a web account? Register with us today!
18084985
View Documents
Novus Biologicals™
NBP1-50840-0.01MG
0.01 mg
403.00€
0.01mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 21-119 of Human beta 2-Microglobulin The Recombinant Human beta 2-Microglobulin Protein is derived from E. coli. The Recombinant Human beta 2-Microglobulin Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

0.5mg/mL
Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 20% glycerol, 2mM DTT, 100mM NaCl
SDS-PAGE
RUO
Unconjugated
SDS-PAGE
14.0kDa
Store at -80°C. Avoid freeze-thaw cycles.
beta 2-Microglobulin
>95%
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Novus Biologicals™ beta 2-Microglobulin Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.