missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ TBE Gels, 6%

Novex™ TBE Gels 6% are designed to run on the XCell SureLock™ Mini-Cell and provide high-resolution analysis of restriction digests and PCR products.

Brand:  Invitrogen™ EC62652BOX

156.60 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 12005735

  • 261.00€ / Pack of 10
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket

Includes: Catalog numbers for gels refer to a single box. One box contains 10 gels.
This item is not returnable. View return policy

Description

Description

Novex™ TBE Gels 6% provide high-resolution analysis of restriction digests and PCR products. Designed to run on the XCell SureLock™ Mini-Cell, the polyacrylamide gels give sharp, clearly resolved, intense bands, and provide separations of double-strand DNA fragments from 65–250 bp.

Novex TBE Gels are available in a variety of well formats and gel percentages and can be stained by silver staining, ethidium­ bromide, and SYBR™ Green staining techniques after electrophoresis.

Advantages of TBE Gels for nucleic acid separation:
• High resolution and sensitivity with lower background staining
• Requires ∼10% sample concentration and volume of large or agarose gels
• Efficient blotting
• Easy extraction of DNA from gels
• Gel extraction does not interfere with enzymatic reactions
• Accurate and reproducible results

Formulation
Novex TBE gels are manufactured with high-purity Tris base, boric acid, EDTA, acrylamide, bisacrylamide, TEMED, and APS.

Recommended buffers
For optimum performance, Novex™ TBE Running Buffer and Novex™ Hi-Density TBE Sample Buffer are strongly recommended for use with these gels. Novex Hi-Density TBE Sample Buffer contains the tracking dyes Bromophenol Blue and Xylene Cyanol as well as the density agent Ficoll™, which yields sharper and straighter bands than conventional density agents.

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• Catalog number refers to a single box of 10 gels

Store at 4°C.
XCell SureLock Mini-Cell
TBE
65 to 250 bp
10 Gels/Box
Wet Ice
Novex TBE Gels, 6%, 12-well
6%
TBE Gel
12-well
Double-stranded DNA (dsDNA)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions