missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ Large Protein Staining Kit

Specifically designed for large protein analysis on NuPAGE Novex Tris-Acetate Gels under denaturing conditions

Brand:  Invitrogen™ LP0002

Product Code. 10626163

  • 972.00€ / 1 set
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The HiMark™ Pre-Stained and Unstained High Molecular Weight Protein Standards offer a time-saving load-and-go format. The HiMark Pre-stained Standard includes one pink colored orientation band and eight blue bands ranging in apparent molecular weight of 31 - 460 kDa in NuPAGE Tris-Acetate Gel/SDS buffer system. It's suitable for monitoring electrophoresis, western transfer, and approximating molecular weight of unknown big proteins. The HiMark Unstained Standard, along with the NuPAGE Tris-Acetate Gel/SDS buffer system, offers the highest accuracy for molecular weight estimation of large proteins. It consists of nine proteins, ranging in size from 40 to 500 kDa in NuPAGE Novex 3-8% and 7% Tris-Acetate Gel/SDS buffer systems. Visualize with Coomassie™ stain, silver, and fluorescent protein stains post-electrophoresis. Several NuPAGE Large Protein Starter Kits are available to provide you with all necessary reagents and optimized protocols for high-resolution large protein separation and analysis. Each kit includes two boxes of NuPAGE 3-8% Tris-Acetate Gels, Tris-Acetate SDS Buffer Kit, a staining reagent or blotting membranes, and a vial of the appropriate HiMark Standard.

  • Gel Percentage: 3-8 %
  • Gel Size: Mini (8 cm x 8 cm)
  • Well Format: 1D Well

1D Gel Electrophoresis, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

TRUSTED_SUSTAINABILITY
Specifications

Specifications

See product manual for kit contents and component storage conditions. Components of each kit are also available individually.
3 to 8%
Tris-Acetate
25 μL
Unstained
NuPAGE™
1.0 mm
XCell SureLock Mini-Cell
Mini
Protein Stain Kit
40 to 500 kDa
10-well
20 gels
1D Well
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.