Learn More
Invitrogen™ Novex™ Midi Tris-Glycine Plus Welcome Pack, 4 to 12%
The Novex Tris-Glycine Plus Midi Welcome Pack provides all of the necessary Novex Tris-Glycine Plus Midi gels, buffers, and reagents needed to begin using the SureLock Tandem Midi Gel Tank.
756.00€
Specifications
For Use With (Equipment) | SureLock Tandem Midi Gel Tank |
---|---|
Gel Percentage | 4 to 12% |
Gel Size | Midi |
Gel Thickness | 1.0 mm |
Gel Type | Tris-Glycine |
Product Code | Brand | Wells | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Wells | Price | Quantity & Availability | |||||
16686926
|
Invitrogen™
STM4010 |
20-well |
756.00€
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
17144619
|
Invitrogen™
STM4011 |
26-well |
756.00€
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||||
Description
The Novex Tris-Glycine Plus Midi Welcome Pack provides all of the necessary Novex Tris-Glycine Plus Midi gels, buffers, and reagents you will need to begin using the SureLock Tandem Midi Gel Tank. The SureLock Tandem Midi Gel Tank provides for rapid electrophoresis of midi gels using minimal buffer (∼520 mL/gel) in a leak-free system. With a set-up time of ∼30 seconds, the tank efficiently runs midi gels while also providing consistent performance. The tank features two independent chambers, allowing electrophoresis of one or two gels at a time, which saves on buffer and limits waste.
The Novex Tris-Glycine Plus Midi Welcome Pack contains:
• SureLock Tandem Midi Gel Tank (STM1001)
• Novex Tris-Glycine Plus Midi Gels (2 boxes, 20 gels)
• Novex Tris-Glycine SDS Running Buffer, 10X (LC2675)
• Novex Tris-Glycine SDS Sample Buffer, 2X (LC2676)
• NuPAGE Sample Reducing Agent, 10X (NP0004)
• PageRuler Plus Prestained Protein Ladder, 10 to 250 kDa (26619)
About the SureLock Tandem Midi Gel Tank
The SureLock Tandem Midi Gel Tank provides for rapid electrophoresis of midi gels using minimal buffer (∼520 mL/gel) in a leak-free system. With a set-up time of ∼30 seconds, the tank efficiently runs midi gels while also providing consistent performance. The tank features two independent chambers, allowing electrophoresis of one or two gels at a time, which saves on buffer and limits waste. The tank is compatible with all Invitrogen pre-cast midi gels and midi gel cassettes.
About Novex Tris-Glycine Plus Midi Gels
Novex Tris-Glycine Plus midi gels are polyacrylamide gels based on traditional Laemmli protein electrophoresis, allowing the use of Laemmli sample and running buffers. The gels offer reproducible separation of a wide range of proteins into well-resolved bands. Novex Tris-Glycine Plus Midi gels are a 1.0-mm thick, wider format (8 cm x 13 cm) gel for higher throughput electrophoresis of protein samples.
Transfer and detection
For transfer of proteins to a membrane, we recommend using Novex Tris-Glycine Transfer Buffer (LC3675) for traditional wet transfer using the SureLock Tandem Midi Blot Module (STM2001). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).
Specifications
SureLock Tandem Midi Gel Tank | |
Midi | |
Tris-Glycine | |
1 Welcome Pk. |
4 to 12% | |
1.0 mm | |
NOVEX™ | |
Wet Ice |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.