Learn More
Laboratoires Anios™ Non-Autoclavable Instrubac, Polypropylene
Laboratoires Anios™ Non-Autoclavable Instrubac, Polypropylene is a large-volume container with better mechanical resistance and can be used as a soaking bin for the collection of stained instrumentation.
Brand:  Laboratoires Anios™ 404412
Product Code. 15765643
  • 877.00€ / Each
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Description
  • Soaking bin
  • Non-autoclavable
  • With tap
Specifications
Specifications
Soaking Bin
40 L
Tap
51 cm
22 cm
Polypropylene
Not Autoclavable
51 x 34 x 22 cm
34 cm