missing translation for 'onlineSavingsMsg'
Learn More

NNC 711, Tocris Bioscience™

Product Code. 18084582
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective GAT-1 inhibitor

Potent and selective inhibitor of GABA uptake by GAT-1 (IC50 values are 0.04, 171, 1700 and 622 μM for hGAT-1, rGAT-2, hGAT-3 and hBGT-1 respectively). Anticonvulsant following systemic administration in vivo. Also exhibits cognition-enhancing activity.

Chemical Identifiers

CAS 145645-62-1
Molecular Formula C21H23ClN2O3
Molecular Weight (g/mol) 386.876
InChI Key YZYRTEYMUTWJPL-UHFFFAOYSA-N
Synonym no-711 hydrochloride, 1-2-diphenylmethylene amino oxy ethyl-1,2,5,6-tetrahydro-3-pyridinecarboxylic acid, 1,2,5,6-tetrahydro-1-2-diphenylmethylene amino oxy ethyl-3-pyridinecarboxylic acid hydrochloride, 1-2-diphenylmethylene imino oxy ethyl-1,2,5,6-tetrahydro-3-pyridinecarboxylic acid hydrochloride hydrochloride, 3-pyridinecarboxylic acid, 1,2,5,6-tetrahydro-1-2-diphenylmethylene amino oxy ethyl-, monohydrochloride, c21h22n2o3.hcl, no-711 hydrochloride hplc, 1-2-diphenylmethylene imino oxy ethyl-1,2,5,6-tetrahydro-3-pyridine-carboxylic acid hydrochloride, 1-2-diphenylmethylidene amino oxy ethyl-5,6-dihydro-2h-pyridine-3-carboxylic acid hydrochloride, 1-2-benzhydrylideneamino oxyethyl-3,6-dihydro-2h-pyridine-5-carboxylic acid hydrochloride
PubChem CID 123738
IUPAC Name 1-[2-(benzhydrylideneamino)oxyethyl]-3,6-dihydro-2H-pyridine-5-carboxylic acid;hydrochloride
SMILES C1CN(CC(=C1)C(=O)O)CCON=C(C2=CC=CC=C2)C3=CC=CC=C3.Cl

Specifications

Quantity 10 mg
Formula Weight 386.88
Percent Purity >99%
Chemical Name or Material NNC 711
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.