missing translation for 'onlineSavingsMsg'
Learn More

NNC 63-0532, Tocris Bioscience™

Product Code. 30001286
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent non-peptide NOP agonist; brain penetrant

Potent non-peptide agonist for the NOP receptor (EC50 = 305 nM, Ki = 7.3 nM); ∽ 12-fold selective over other receptors. Centrally active following systemic administration in vivo.

Chemical Identifiers

CAS 250685-44-0
Molecular Formula C27H29N3O3
Synonym 8-(1-Naphthalenylmethyl)-4-oxo-1-phenyl-1,3,8-triazaspiro[4.5]decane-3-acetic acid, methyl ester

Specifications

Quantity 10 mg
Formula Weight 443.54
Percent Purity >98%
Chemical Name or Material NNC 63-0532
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'