Learn More
Schmidlin™ NM Plus Rack Hydrogen Generator
This generator produces a continuous supply of high purity, dry hydrogen.
Brand: Schmidlin™ DB-RNM500-EU
This item is not returnable.
View return policy
Description
- Increase efficiency and improve safety and performance
- On-demand supply 24/7
- Pressure: 11 barg (160 psig)
- Unique permeation membrane drying system
- Cold dual dynamic regeneration dryer
- USB connectivity
- Easy to install with complete 'plug and play' functionality
- Ideal for all GC detector applications
- Meets and exceeds requirements for the most demanding GC applications
- Superior hydrogen production with reliable long life cell
- Minimal maintenance; no desiccant cartridges to change
- PC monitoring for maintenance, diagnostics and remote connection
Specifications
500 cm | |
13.3 cm | |
11 bar | |
48.5 cm | |
>99.99996% |
500 mL/min. | |
35°C | |
19 in. | |
Outlet: 1/8 in. | |
200 W |