missing translation for 'onlineSavingsMsg'
Learn More

Ansell NITROTOUGH™ N630 Heavy Duty Nitrile-Coated Gloves - Gauntlet Cuff

Product Code. p-7215036 Shop All Ansell Products
Click to view available options
Size:
8
9
10
11
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 16855320

Brand: Ansell C329080

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Nitrile coating with support provides an effective barrier against grease and oil without degrading like leather or cotton. Gloves feature a 100% cotton fleece interlock liner for increased user comfort.

  • Nitrile coating covers the fingers and the knuckles for an effective barrier against oil and grease
  • 100% cotton fleeced interlock liner for increased comfort
  • Stronger and more flexible than PVC
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 6
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material Nitrile
Color Blue
Cut Resistance Rating ANSI A2
Hand Specific
Size 8
EN388 Standard 4221B
For Use With (Application) Automotive Assembly and Metal Fabrication
Glove Cuff Color Natural
Glove Cuff Style Safety
Liner Material 100% Cotton
Series N630
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.