Learn More
WTW™ Nitrite Reagent Test Kits
With certificate for optimum precision.
Brand:  WTW™ 252069
Product Code. 12055615
  • 159.00€ / Pack of 25
Estimated Shipment: 28-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Convenient and cost-effective
  • Assured quality by AQA/IQC
  • For low concentrations and routine work with large sample throughput in the laboratory
  • Wide measuring range using rectangular cuvettes for determination of trace concentrations
TRUSTED_SUSTAINABILITY
Specifications
Specifications
25
3.3 - 295.2mg/L
25 Tests
Nitrite (NO2) reagent test kit
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • nitrite cell test method
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H332-Harmful if inhaled.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : dipotassium disulphate