missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric acid, 90+%, ACS reagent, fuming, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Blue to Blue-Green |
CAS | 7697-37-2 |
Melting Point | -40.0°C |
Boiling Point | 84.0°C |
Density | 1.5110g/mL |
Heavy Metals (as Pb) | 5ppm max. |
Packaging | Glass bottle |
Residue after Ignition | 0.002 % max. |
Specific Gravity | 1.511 |
Assay Percent Range | 90+% |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0466855 | Certificate of Analysis | 06/11/2024 | 270620010 |
A0457056 | Certificate of Analysis | 28/10/2024 | 270620010 |
A0452742 | Certificate of Analysis | 02/05/2024 | 270620010 |
A0450266 | Certificate of Analysis | 12/02/2024 | 270620010 |
A0425777 | Certificate of Analysis | 08/01/2024 | 270620010 |
Safety and Handling



Product Identifier
- Nitric acid fuming, 85-100%
- Danger
- Acute toxicity Category 1
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Oxidising liquid Category 3
- Skin corrosion/irritation Category 1 A
- H272-May intensify fire; oxidiser.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H330-Fatal if inhaled.
- EUH071-Corrosive to the respiratory tract.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Nitric acid
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Nitric acid, 90+%, ACS reagent, fuming, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction