missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Nitrate Reagent Sets, For Colorimetric Water Testing

Product Code. 11728763
Click to view available options
Quantity:
1 Set
Unit Size:
Pack of 50
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 11728763

Brand: Hanna Instruments™ HI9376650

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Nitrate Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured solutions for use with compatible benchtop and/or portable Hanna meters.

These high quality reagents are designed to follow accepted test methods for colorimetric determination of nitrates. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.

  • Reagent sets available with ready-to-use vials (chromotropic method) or individual reagent packets (cadmium reduction method)
  • Pre-made, pre-measured water testing reagents for ease of use
  • Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
  • Clearly marked with expiration date and lot number on each packet and bottle for traceability

Specifications

Analytes Chromotropic Acid
For Use With (Application) Nitrate determination
For Use With (Equipment) Compatible Hanna benchtop and portable photometers
Includes Ready-to-use vials (HI93766V-0) and Individual packets (HI93766-0)
No. of Tests 50
Quantity 1 Set
Range 0.0 to 30.0mg/L as NO3-N
Test Method Cholorimetric determination
Type Water Testing Kit
compliance-icons
Product Identifier
  • X50 NITRATE REAGENT 0630 MG/L
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : chromotropic acid disodium salt, sodiummetabisulphate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.