missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nisoxetine hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
A potent and selective inhibitor of noradrenalin uptake with little or no affinity for a range of other neurotransmitter receptors.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 307.82 |
Percent Purity | >99% |
Chemical Name or Material | Nisoxetine hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'