missing translation for 'onlineSavingsMsg'
Learn More

Nicotinamide Adenine Dinucleotide, Serva Electrophoresis™

Product Code. 11458462
Encompass_Preferred
Click to view available options
Quantity:
1 g
Packaging:
1g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 53-84-9
Packaging 1g
Molecular Formula C21H27N7O14P2
MDL Number MFCD00150377
Quantity 1 g
Synonym nadide, nad+, coenzyme i, beta-nad, beta-nad+, diphosphopyridine nucleotide, beta-diphosphopyridine nucleotide, enzopride, nicotinamide adenine dinucleotide, codehydrase i
InChI Key BAWFJGJZGIEFAR-WIWLTUSXNA-N
SMILES NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
IUPAC Name 1-[(2R,3R,4S,5R)-5-{[({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]methyl}-3,4-dihydroxyoxolan-2-yl]-3-carbamoyl-1λ⁵-pyridin-1-ylium
Molecular Weight (g/mol) 663.43
PubChem CID 15938971
ChEBI CHEBI:57540
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'