missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Nickel Cell Test Kit

With certificate for optimum precision.

Brand:  WTW™ 250409


Product Code. 11724774

  • 147.00€ / 0.11kg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Convenient and cost-effective
  • Assured quality by AQA/IQC
  • For infrequent or very large sample throughput in the laboratory
  • Sample adding, inserting, measuring and reading
  • Reaction cuvette with barcode or method selection

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dimethylglyoxime
PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers
25
25 Tests
CombiCheck Nickel (Ni) cell test kit
For waste water, drinking water, surface water
Certificates
0.10 - 6.00mg/I Ni
Photometric
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Nickel Cell Test Method: photometric 0.10 - 6.00 mg/l Ni Spectroquant

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Flammable liquid Category 2

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : ammonia solution

SDS
Documents

Documents

Certificates
Special Offers

Special Offers