Learn More
WTW™ Nickel Reagent Test Kit
With certificate for optimum precision.
Brand:  WTW™ 250443
Product Code. 10603562
  • 203.00€ / Each
Explore more special offers
Description
Description
  • Convenient and cost-effective
  • Assured quality by AQA/IQC
  • For low concentrations and routine work with large sample throughput in the laboratory
  • Wide measuring range using rectangular cuvettes for determination of trace concentrations
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Barbituric acid-pyridine Carboxylic Acid
For PhotoLab™ S12, PhotoLab™ Spectral, spectroFlex photometers
250
Photometric
For waste water, drinking water, surface water
Certificates
0.02 - 5.00mg/I Ni
Photometric test kit
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • nickel test method
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : iodine